Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016170.4 | NP_057254.1 | 2 | 956 | ||
GenBank mRNA | AB008501.1 | - | 2 | 618 | ||
AB593086.1 | - | 3 | 643 | |||
BC006356.2 | - | 2 | 941 | 4125397 | ||
BC006396.1 | - | 1 | 1213 | |||
BC018611.1 | - | 3 | 552 |