Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018951.3 | NP_061824.3 | 2 | 1240 | ||
NR_037939.1 | 2 | 868 | ||||
GenBank mRNA | AK312450.1 | - | 2 | 660 | ||
BC013971.2 | - | 2 | 1208 | 4107013 | ||
BC071843.1 | - | 2 | 503 | |||
BC094807.1 | - | 2 | 503 |
Gene Symbol: HOXA10-HOXA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FY213345.1 | - | 2 | 381 |