Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004503.3 | NP_004494.1 | 1 | 366 | ||
NM_153693.4 | NP_710160.1 | 2 | 763 | |||
GenBank mRNA | AK314829.1 | - | 2 | 443 | ||
AW445186.1 | - | 2 | 177 | |||
BC074844.2 | - | 1 | 280 | 30915460 | ||
BC074845.2 | - | 1 | 280 | 30915223 | ||
CR456954.1 | - | 1 | 254 | |||
M16938.1 | - | 2 | 490 | |||
S82986.1 | - | 1 | 386 |