Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021193.3 | NP_067016.3 | 1 | 397 | ||
GenBank mRNA | BC121103.1 | - | 1 | 401 | 40120251 | |
BC121104.1 | - | 1 | 401 | 40120253 |