Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258357.1 | NP_001245286.1 | 3 | 211 | ||
NM_001258358.1 | NP_001245287.1 | 3 | 223 | |||
NM_001258359.1 | NP_001245288.1 | 3 | 617 | |||
NM_002149.3 | NP_002140.2 | 3 | 437 | |||
NM_134421.2 | NP_602293.1 | 4 | 579 | |||
XM_005246160.1 | XP_005246217.1 | 4 | 502 | |||
XM_005246161.1 | XP_005246218.1 | 4 | 625 | |||
XM_005246162.1 | XP_005246219.1 | 4 | 628 | |||
XM_005246163.1 | XP_005246220.1 | 5 | 675 | |||
XM_011510346.2 | XP_011508648.1 | 3 | 281 | |||
XM_011510347.1 | XP_011508649.1 | 5 | 722 | |||
XM_011510348.1 | XP_011508650.1 | 4 | 515 | |||
XM_011510349.1 | XP_011508651.1 | 3 | 452 | |||
XM_017003950.1 | XP_016859439.1 | 5 | 2169 | |||
XM_017003951.1 | XP_016859440.1 | 5 | 652 | |||
XM_017003952.1 | XP_016859441.1 | 3 | 397 | |||
XM_017003953.1 | XP_016859442.1 | 4 | 532 | |||
GenBank mRNA | AK000596.1 | - | 4 | 585 | ||
AK292280.1 | - | 3 | 617 | |||
BC009846.2 | - | 3 | 412 | 4100091 | ||
BC017028.1 | - | 3 | 404 | 3902811 | ||
BC017482.1 | - | 3 | 427 | 3504942 | ||
BI601950.1 | - | 3 | 228 | |||
BI603158.1 | - | 3 | 442 | |||
BT009835.1 | - | 1 | 53 | |||
BU145561.1 | - | 3 | 218 | |||
CR541975.1 | - | 1 | 53 | |||
CR749837.1 | - | 5 | 552 |