Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPCAL1 SIRNA Assays ›
Gene Symbol:
HPCAL1
Gene Name:
hippocalcin like 1
Gene Aliases:
BDR1, HLP2, VILIP-3
Chromosome Location:
Chr.2: 10302124 - 10427617 on Build GRCh38
UniGene ID:
Hs.580427
Species:
Human
siRNA ID
HSS142532
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPCAL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258357.1
NP_001245286.1
3, 4
546
NM_001258358.1
NP_001245287.1
3, 4
558
NM_001258359.1
NP_001245288.1
3, 4
952
NM_002149.3
NP_002140.2
3, 4
772
NM_134421.2
NP_602293.1
4, 5
914
XM_005246160.1
XP_005246217.1
4, 5
837
XM_005246161.1
XP_005246218.1
4, 5
960
XM_005246162.1
XP_005246219.1
4, 5
963
XM_005246163.1
XP_005246220.1
5, 6
1010
XM_011510346.2
XP_011508648.1
3, 4
616
XM_011510347.1
XP_011508649.1
5, 6
1057
XM_011510348.1
XP_011508650.1
4, 5
850
XM_011510349.1
XP_011508651.1
3, 4
787
XM_017003950.1
XP_016859439.1
5, 6
2504
XM_017003951.1
XP_016859440.1
5, 6
987
XM_017003952.1
XP_016859441.1
3, 4
732
XM_017003953.1
XP_016859442.1
4, 5
867
GenBank mRNA
AF070616.1
-
1, 2
141
AK000596.1
-
4, 5
920
AK292280.1
-
3, 4
952
BC009846.2
-
3, 4
747
4100091
BC017028.1
-
3, 4
739
3902811
BC017482.1
-
3, 4
762
3504942
BI601950.1
-
3, 4
563
BT009835.1
-
1, 2
388
BU145561.1
-
3, 4
553
CR541975.1
-
1, 2
388
CR749837.1
-
5, 6
887
D16227.1
-
2, 3
535
Back To Top
More Information
Gene Ontology Categories:
Process(es)
calcium ion binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-zpscg:80/10.239.160.225:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0