Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002159.3 | NP_002150.1 | 5 | 215 | ||
GenBank mRNA | BC017835.1 | - | 5 | 188 | 4289874 | |
BP335073.2 | - | 5 | 192 | |||
CB984538.1 | - | 5 | 219 | |||
CR627329.1 | - | 3 | 395 | |||
M26664.1 | - | 4 | 130 |