Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001546.3 | NP_001537.1 | 1 | 682 | ||
GenBank mRNA | BC014941.1 | - | 1 | 666 | 4552357 | |
U16153.1 | - | 1 | 593 | |||
U28368.1 | - | 1 | 672 | |||
Y07958.1 | - | 1 | 332 |