Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000879.2 | NP_000870.1 | 1 | 85 | ||
XM_005271988.3 | XP_005272045.1 | 2 | 219 | |||
XM_011543373.2 | XP_011541675.1 | 3 | 321 | |||
XM_011543374.2 | XP_011541676.1 | 2 | 272 | |||
GenBank mRNA | BC066282.1 | - | 1 | 64 | 6971771 | |
DQ655974.2 | - | 1 | 484 | |||
X04688.1 | - | 1 | 85 | |||
X12705.1 | - | 1 | 85 |