Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001572.3 | NP_001563.2 | 9 | 1297 | ||
NM_004029.2 | NP_004020.1 | 8 | 1210 | |||
NM_004031.2 | NP_004022.2 | 7 | 1376 | |||
XM_005252906.3 | XP_005252963.1 | 8 | 1362 | |||
XM_005252907.3 | XP_005252964.1 | 8 | 1359 | |||
XM_005252909.3 | XP_005252966.1 | 7 | 1275 | |||
XM_011520066.2 | XP_011518368.1 | 9 | 1273 | |||
XM_017017674.1 | XP_016873163.1 | 2 | 175 | |||
GenBank mRNA | AF076494.1 | - | 7 | 1301 | ||
AK303752.1 | - | 7 | 1108 | |||
BC136555.1 | - | 7 | 1012 | 9020545 | ||
KC477210.1 | - | 6 | 1289 | |||
U53830.1 | - | 8 | 1219 | |||
U53831.1 | - | 7 | 1132 | |||
U53832.1 | - | 7 | 1051 | |||
U73036.1 | - | 9 | 1236 |