Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001572.3 | NP_001563.2 | 11 | 1828 | ||
NM_004029.2 | NP_004020.1 | 10 | 1741 | |||
NM_004031.2 | NP_004022.2 | 9 | 1907 | |||
XM_005252906.3 | XP_005252963.1 | 10 | 1893 | |||
XM_005252907.3 | XP_005252964.1 | 10 | 1890 | |||
XM_005252909.3 | XP_005252966.1 | 9 | 1806 | |||
XM_011520066.2 | XP_011518368.1 | 11 | 1804 | |||
XM_017017674.1 | XP_016873163.1 | 4 | 706 | |||
GenBank mRNA | AF076494.1 | - | 9 | 1832 | ||
AK303752.1 | - | 9 | 1639 | |||
BC136555.1 | - | 9 | 1543 | 9020545 | ||
KC477210.1 | - | 8 | 1820 | |||
U53830.1 | - | 10 | 1750 | |||
U53831.1 | - | 9 | 1663 | |||
U53832.1 | - | 9 | 1582 | |||
U73036.1 | - | 11 | 1767 |