Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039574.2 | NP_001034663.1 | 2 | 1616 | ||
NM_004978.4 | NP_004969.2 | 2 | 1616 | |||
NR_036437.1 | 2 | 1616 | ||||
XM_006710625.3 | XP_006710688.1 | 2 | 1460 | |||
XM_006710627.3 | XP_006710690.1 | 2 | 1460 | |||
XM_011541401.2 | XP_011539703.1 | 2 | 1460 | |||
GenBank mRNA | AK310193.1 | - | 2 | 1041 | ||
BC101769.1 | - | 2 | 887 | 8069275 | ||
BX537638.1 | - | 1 | 3404 | |||
M64676.1 | - | 2 | 986 |