Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005583.4 | NP_005574.2 | 4 | 1463 | ||
XM_005259912.3 | XP_005259969.1 | 4 | 1301 | |||
XM_017026816.1 | XP_016882305.1 | 5 | 2163 | |||
XM_017026817.1 | XP_016882306.1 | 5 | 2016 | |||
GenBank mRNA | AI554151.1 | - | 1 | 323 | ||
BC002796.2 | - | 4 | 1170 | 3619444 | ||
M22637.1 | - | 4 | 1028 |