Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199216.1 | NP_001186145.1 | 7 | 1400 | ||
NM_002361.3 | NP_002352.1 | 7 | 1413 | |||
NM_080600.2 | NP_542167.1 | 7 | 1413 | |||
GenBank mRNA | AK094545.1 | - | 8 | 1259 | ||
AK222649.1 | - | 7 | 1338 | |||
AK222680.1 | - | 7 | 1316 | |||
AK223562.1 | - | 7 | 1374 | |||
AK308976.1 | - | 7 | 1325 | |||
BC093045.1 | - | 7 | 1365 | 30396005 | ||
M29273.1 | - | 7 | 1335 |