Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
siRNA ID
HSS142867
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
8
926
NM_002374.3
NP_002365.3
10
4791
NM_031845.2
NP_114033.2
7
723
NM_031847.2
NP_114035.2
7
723
XM_005246555.1
XP_005246612.1
11
5156
XM_005246565.1
XP_005246622.1
7
4445
XM_005246566.1
XP_005246623.1
7
4433
XM_006712532.1
XP_006712595.1
11
5153
XM_011511186.1
XP_011509488.1
11
5063
XM_011511187.1
XP_011509489.1
10
4957
XM_011511188.1
XP_011509490.1
10
4957
XM_011511190.1
XP_011509492.1
10
4960
XM_011511191.1
XP_011509493.1
9
4825
XM_011511192.1
XP_011509494.1
10
4960
XM_011511193.1
XP_011509495.1
10
4957
XM_011511195.1
XP_011509497.1
9
4711
XM_011511196.2
XP_011509498.1
10
4898
XM_011511197.2
XP_011509499.1
10
4895
XM_017004111.1
XP_016859600.1
10
4957
XM_017004112.1
XP_016859601.1
10
4910
XM_017004113.1
XP_016859602.1
10
4802
XM_017004114.1
XP_016859603.1
10
4910
XM_017004115.1
XP_016859604.1
9
4702
XM_017004116.1
XP_016859605.1
9
4699
XM_017004117.1
XP_016859606.1
9
4699
XM_017004118.1
XP_016859607.1
10
4817
XM_017004119.1
XP_016859608.1
10
4907
XM_017004120.1
XP_016859609.1
10
4907
XM_017004121.1
XP_016859610.1
10
4814
XM_017004122.1
XP_016859611.1
10
4814
XM_017004123.1
XP_016859612.1
10
4898
XM_017004124.1
XP_016859613.1
10
4805
XM_017004125.1
XP_016859614.1
10
4895
XM_017004126.1
XP_016859615.1
10
4895
XM_017004127.1
XP_016859616.1
10
4895
XM_017004128.1
XP_016859617.1
9
4696
XM_017004129.1
XP_016859618.1
9
4693
XM_017004130.1
XP_016859619.1
7
4882
XM_017004131.1
XP_016859620.1
7
4445
XM_017004132.1
XP_016859621.1
7
4433
XM_017004133.1
XP_016859622.1
7
4445
XM_017004134.1
XP_016859623.1
7
4433
XM_017004135.1
XP_016859624.1
7
4879
XM_017004136.1
XP_016859625.1
7
4442
XM_017004137.1
XP_016859626.1
7
4430
XM_017004138.1
XP_016859627.1
7
749
XM_017004139.1
XP_016859628.1
4
506
XM_017004140.1
XP_016859629.1
7
749
GenBank mRNA
AB209330.1
-
6
641
AK291446.1
-
4
512
BC027583.1
-
6
624
BC038857.1
-
8
915
BC110423.1
-
7
971
BC117123.1
-
5
552
BC143245.1
-
5
557
M25668.1
-
4
3064
U01828.1
-
10
4791
U89329.1
-
4
475
U89330.1
-
7
4531
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-n26sq:80/10.239.164.94:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline