Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005368.2 | NP_005359.1 | 2, 3 | 396 | ||
NM_203377.1 | NP_976311.1 | 3, 4 | 488 | |||
NM_203378.1 | NP_976312.1 | 3, 4 | 471 | |||
XM_005261605.2 | XP_005261662.1 | 3, 4 | 432 | |||
GenBank mRNA | AK300657.1 | - | 3, 4 | 354 | ||
AK314057.1 | - | 3, 4 | 457 | |||
BC014547.1 | - | 2, 3 | 392 | 4244838 | ||
BC018001.1 | - | 2, 3 | 361 | 4271145 | ||
BQ925917.1 | - | 520 | ||||
BQ956082.1 | - | 375 | ||||
BU585249.1 | - | 2, 3 | 517 | |||
CB140824.1 | - | 3, 4 | 501 | |||
CR456516.1 | - | 2, 3 | 364 | |||
CR541949.1 | - | 2, 3 | 316 |