Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL1 SIRNA Assays ›
Gene Symbol:
MBNL1
Gene Name:
muscleblind like splicing regulator 1
Gene Aliases:
EXP, MBNL
Chromosome Location:
Chr.3: 152243656 - 152465780 on Build GRCh38
UniGene ID:
Hs.201858
Species:
Human
siRNA ID
HSS142883
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001314057.1
NP_001300986.1
4
870
NM_021038.4
NP_066368.2
3
1655
NM_207292.2
NP_997175.1
3
1655
NM_207293.1
NP_997176.1
2
1030
NM_207294.1
NP_997177.1
2
1030
NM_207295.1
NP_997178.1
2
1030
NM_207296.1
NP_997179.1
2
1030
NM_207297.1
NP_997180.1
2
1030
XM_005247457.4
XP_005247514.1
4
1223
XM_005247458.4
XP_005247515.1
4
2477
XM_005247460.4
XP_005247517.1
4
1186
XM_005247461.4
XP_005247518.1
4
1223
XM_005247462.4
XP_005247519.1
4
1223
XM_005247463.4
XP_005247520.1
4
1223
XM_005247464.4
XP_005247521.1
3
1281
XM_005247465.4
XP_005247522.1
4
1223
XM_005247466.4
XP_005247523.1
3
1258
XM_005247467.4
XP_005247524.1
4
1223
XM_005247468.4
XP_005247525.1
4
1223
XM_005247471.4
XP_005247528.1
3
1253
XM_005247474.3
XP_005247531.1
2
141
XM_005247475.3
XP_005247532.1
3
464
XM_005247476.4
XP_005247533.1
2
231
XM_005247477.4
XP_005247534.1
2
230
XM_006713639.1
XP_006713702.1
4
1205
XM_011512843.2
XP_011511145.1
4
1223
XM_011512845.2
XP_011511147.1
4
1223
XM_011512846.2
XP_011511148.1
4
1223
XM_011512847.2
XP_011511149.1
3
1186
XM_011512848.2
XP_011511150.1
4
827
XM_011512849.2
XP_011511151.1
3
1130
XM_017006423.1
XP_016861912.1
3
1474
XM_017006424.1
XP_016861913.1
3
1301
XM_017006425.1
XP_016861914.1
3
1303
XM_017006426.1
XP_016861915.1
3
1301
XM_017006427.1
XP_016861916.1
3
1448
XM_017006428.1
XP_016861917.1
3
1112
XM_017006429.1
XP_016861918.1
3
1195
XM_017006430.1
XP_016861919.1
3
1259
XM_017006431.1
XP_016861920.1
3
1336
XM_017006432.1
XP_016861921.1
3
1258
XM_017006433.1
XP_016861922.1
3
1258
XM_017006434.1
XP_016861923.1
3
1264
XM_017006435.1
XP_016861924.1
3
271
XM_017006436.1
XP_016861925.1
3
2384
XM_017006437.1
XP_016861926.1
3
1439
XM_017006438.1
XP_016861927.1
3
1130
XM_017006439.1
XP_016861928.1
3
464
XM_017006440.1
XP_016861929.1
2
198
XM_017006441.1
XP_016861930.1
2
231
XM_017006442.1
XP_016861931.1
2
334
XM_017006443.1
XP_016861932.1
2
342
GenBank mRNA
AB007888.1
-
3
1655
AF255334.1
-
2
752
AF395876.1
-
2
241
AF401998.1
-
2
241
AF497718.1
-
2
241
AF497719.1
-
2
241
AJ308400.1
-
2
270
BC043493.1
-
3
1160
6044671
BC050535.1
-
4
391
6042311
Y13829.1
-
2
752
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
mRNA processing
nervous system development
RNA splicing
embryonic limb morphogenesis
regulation of RNA splicing
myoblast differentiation
regulatory region RNA binding
RNA binding
double-stranded RNA binding
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0