Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030663.2 | NP_109588.2 | 2 | 352 | ||
GenBank mRNA | AK312020.1 | - | 2 | 352 | ||
BC014593.2 | - | 2 | 352 | 4824172 | ||
BC016744.2 | - | 2 | 335 | 4829283 | ||
X89960.1 | - | 2 | 332 |