Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318933.1 | NP_001305862.1 | 22 | 1839 | ||
NM_002390.5 | NP_002381.2 | 22 | 1843 | |||
XM_005257373.3 | XP_005257430.1 | 22 | 1985 | |||
XM_017024647.1 | XP_016880136.1 | 21 | 1757 | |||
GenBank mRNA | AB009675.1 | - | 22 | 1822 | ||
AK296512.1 | - | 22 | 1920 | |||
CN418442.1 | - | 6 | 456 | |||
D17390.1 | - | 19 | 1562 |