Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002415.1 | NP_002406.1 | 1 | 109 | ||
GenBank mRNA | AF469046.1 | - | 1 | 12 | ||
AK311929.1 | - | 1 | 104 | |||
BC000447.2 | - | 1 | 93 | 2821346 | ||
BC007676.1 | - | 1 | 77 | 3605756 | ||
BC008914.2 | - | 1 | 63 | 3615896 | ||
BC013976.2 | - | 1 | 108 | 4130651 | ||
BC022414.1 | - | 1 | 110 | 4243461 | ||
BC053376.1 | - | 1 | 46 | 6047427 | ||
BT007148.1 | - | 1 | 12 | |||
CR407644.1 | - | 1 | 12 | |||
CR456520.1 | - | 1 | 32 | |||
CR541651.1 | - | 1 | 12 | |||
JQ846015.1 | - | 1 | 12 | |||
L10612.1 | - | 1 | 86 | |||
M25639.1 | - | 1 | 62 | |||
Z23063.1 | - | 1 | 12 |
Gene Symbol: MIF-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038911.1 | 3 | 1412 | |||
GenBank mRNA | BC036909.1 | - | 3 | 1412 | ||
DQ595552.1 | - | 11 | ||||
DQ595553.1 | - | 11 |