Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP25-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123723.1 | 3 | 604 | |||
GenBank mRNA | AW204708.1 | - | 3 | 277 | ||
BC026114.2 | - | 2 | 406 | |||
BC045731.1 | - | 2 | 406 | |||
BQ896952.1 | - | 3 | 604 | |||
CN367922.1 | - | 2 | 214 | |||
DA081114.1 | - | 2 | 375 |
Gene Symbol: MMP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023561.1 | XP_016879050.1 | 6 | 1746 | ||
GenBank mRNA | AJ003144.1 | - | 4 | 492 |