Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070236.1 | - | 5 | 1265 | 4290594 | |
U53468.1 | - | 5 | 1278 | |||
U64028.1 | - | 5 | 1271 |