Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278645.1 | NP_001265574.1 | 1 | 142 | ||
NM_001278646.1 | NP_001265575.1 | 1 | 142 | |||
NM_001311168.1 | NP_001298097.1 | 1 | 142 | |||
NM_005005.2 | NP_004996.1 | 1 | 142 | |||
GenBank mRNA | AF044956.1 | - | 1 | 136 | ||
AF067168.1 | - | 1 | 136 | |||
AF261090.1 | - | 1 | 141 | |||
AI907018.1 | - | 1 | 159 | |||
AK302286.1 | - | 1 | 104 | |||
AK313432.1 | - | 1 | 104 | |||
AL136714.1 | - | 1 | 112 | |||
BC007672.2 | - | 1 | 109 | 3160562 | ||
BG703246.1 | - | 1 | 71 | |||
BQ685604.1 | - | 1 | 92 | |||
CN410734.1 | - | 1 | 85 | |||
DB504123.1 | - | 1 | 100 |