Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184986.1 | NP_001171915.1 | 4 | 937 | ||
NM_001184987.1 | NP_001171916.1 | 3 | 405 | |||
NM_001184988.1 | NP_001171917.1 | 4 | 441 | |||
NM_001184989.1 | NP_001171918.1 | 4 | 464 | |||
NM_001184990.1 | NP_001171919.1 | 3 | 401 | |||
NM_001184991.1 | NP_001171920.1 | 3 | 878 | |||
NM_002494.3 | NP_002485.1 | 3 | 540 | |||
GenBank mRNA | AF047184.1 | - | 2 | 135 | ||
AF047435.1 | - | 2 | 115 | |||
AK291147.1 | - | 2 | 145 | |||
BC101798.1 | - | 2 | 151 | 8069304 | ||
BC107682.1 | - | 3 | 510 | 5557684 | ||
BC112061.1 | - | 2 | 151 | 8327529 | ||
BG701467.1 | - | 3 | 544 | |||
BM768825.1 | - | 4 | 441 | |||
BU677336.1 | - | 3 | 279 | |||
CA313899.1 | - | 3 | 316 | |||
CB047240.1 | - | 3 | 326 | |||
CB160001.1 | - | 4 | 392 | |||
CN347371.1 | - | 3 | 306 | |||
CR457005.1 | - | 2 | 107 | |||
W56292.1 | - | 3 | 317 |