Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001503.1 | NP_001001503.1 | 3 | 288 | ||
NM_021075.3 | NP_066553.3 | 4 | 1383 | |||
XM_011529586.2 | XP_011527888.1 | 5 | 1443 | |||
XM_017028359.1 | XP_016883848.1 | 4 | 346 | |||
GenBank mRNA | AK056041.1 | - | 6 | 1840 | ||
AK289586.1 | - | 3 | 242 | |||
AK308228.1 | - | 4 | 1340 | |||
AY173949.1 | - | 3 | 246 | |||
BC021217.2 | - | 4 | 1370 | 4302265 | ||
BC033766.1 | - | 3 | 246 | |||
BC054016.1 | - | 3 | 289 | |||
BU629860.1 | - | 1 | 198 | |||
CR542174.1 | - | 3 | 222 | |||
CR542190.1 | - | 3 | 222 |