Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142393.1 | NP_001135865.1 | 3 | 567 | ||
NM_006403.3 | NP_006394.1 | 2 | 372 | |||
NM_182966.3 | NP_892011.2 | 2 | 372 | |||
NR_073131.1 | 4 | 661 | ||||
GenBank mRNA | AK292682.1 | - | 3 | 567 | ||
BC020686.1 | - | 2 | 343 | |||
BC040207.1 | - | 2 | 358 | 4820745 | ||
BX648041.1 | - | 4 | 629 | |||
DA660812.1 | - | 2 | 373 | |||
DA778416.1 | - | 2 | 372 | |||
L43821.1 | - | 2 | 368 | |||
U64317.1 | - | 2 | 343 |