Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000268.3 | NP_000259.1 | 4 | 843 | ||
NM_016418.5 | NP_057502.2 | 4 | 843 | |||
NM_181825.2 | NP_861546.1 | 4 | 843 | |||
NM_181828.2 | NP_861966.1 | 3 | 717 | |||
NM_181829.2 | NP_861967.1 | 3 | 720 | |||
NM_181830.2 | NP_861968.1 | 2 | 594 | |||
NM_181831.2 | NP_861969.1 | 2 | 594 | |||
NM_181832.2 | NP_861970.1 | 4 | 843 | |||
NM_181833.2 | NP_861971.1 | 4 | 843 | |||
XM_017028809.1 | XP_016884298.1 | 4 | 406 | |||
XM_017028810.1 | XP_016884299.1 | 4 | 402 | |||
GenBank mRNA | AF122827.1 | - | 4 | 450 | ||
AF122828.1 | - | 2 | 201 | |||
AF123570.1 | - | 4 | 450 | |||
AF369657.1 | - | 4 | 825 | |||
AF369658.1 | - | 4 | 825 | |||
AF369661.1 | - | 4 | 400 | |||
AF369662.1 | - | 4 | 400 | |||
AF369663.1 | - | 3 | 274 | |||
AF369664.1 | - | 3 | 277 | |||
AF369665.1 | - | 2 | 151 | |||
AF369666.1 | - | 5 | 516 | |||
AF369667.1 | - | 4 | 425 | |||
AF369668.1 | - | 4 | 400 | |||
AF369701.1 | - | 4 | 766 | |||
AK297116.1 | - | 4 | 782 | |||
BC003112.1 | - | 2 | 423 | 3537045 | ||
BC020257.1 | - | 4 | 738 | 4871980 | ||
CR456530.1 | - | 4 | 462 | |||
L11353.1 | - | 4 | 619 | |||
Z22664.1 | - | 4 | 574 |