Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005598.3 | NP_005589.1 | 2 | 712 | ||
GenBank mRNA | AK313700.1 | - | 2 | 712 | ||
BC013789.1 | - | 2 | 699 | 3866861 | ||
BT006859.1 | - | 1 | 270 | |||
M96739.1 | - | 2 | 714 |