Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322802.1 | NP_001309731.1 | 1 | 153 | ||
NM_005386.3 | NP_005377.1 | 1 | 153 | |||
NM_181689.2 | NP_859017.1 | 1 | 153 | |||
GenBank mRNA | AB002392.1 | - | 1 | 66 | ||
AI879211.1 | - | 1 | 109 | |||
AK312069.1 | - | 1 | 106 | |||
BC001768.1 | - | 1 | 97 | 3353944 | ||
BE255132.1 | - | 1 | 124 | |||
BG472075.1 | - | 1 | 144 | |||
BT019426.1 | - | 1 | 27 | |||
CN388981.1 | - | 1 | 98 | |||
HY021161.1 | - | 1 | 154 | |||
U25033.1 | - | 1 | 96 | |||
U25034.1 | - | 1 | 98 |
Gene Symbol: BLCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167820.1 | NP_001161292.1 | 2 | 229 | ||
XM_017027599.1 | XP_016883088.1 | 1 | 1009 | |||
GenBank mRNA | DA632207.1 | - | 2 | 178 |