Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006191.2 | NP_006182.2 | 6 | 949 | ||
GenBank mRNA | AK291796.1 | - | 6 | 702 | ||
BC001951.1 | - | 6 | 649 | 2823726 | ||
BC007561.1 | - | 6 | 663 | 2988781 | ||
BC019222.1 | - | 7 | 622 | |||
BC032111.2 | - | 6 | 630 | |||
BC069786.1 | - | 6 | 602 | 7262559 | ||
BC072007.1 | - | 6 | 629 | |||
U59435.1 | - | 6 | 627 | |||
U87954.1 | - | 6 | 725 |