Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020872.2 | NP_065923.1 | 2, 3 | 218 | ||
XM_005264757.3 | XP_005264814.1 | 3, 4 | 328 | |||
XM_011533768.2 | XP_011532070.1 | 2, 3 | 3850 | |||
XM_017006507.1 | XP_016861996.1 | 3, 4 | 898 | |||
GenBank mRNA | BC150608.1 | - | 2, 3 | 218 | 9056978 | |
DR002512.1 | - | 2, 3 | 217 |