Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020872.2 | NP_065923.1 | 19 | 2593 | ||
XM_005264757.3 | XP_005264814.1 | 20 | 2703 | |||
XM_011533768.2 | XP_011532070.1 | 19 | 6225 | |||
XM_017006507.1 | XP_016861996.1 | 20 | 3273 | |||
XM_017006508.1 | XP_016861997.1 | 19 | 2498 | |||
XM_017006509.1 | XP_016861998.1 | 10 | 1272 | |||
GenBank mRNA | AB040929.1 | - | 17 | 2242 | ||
BC150608.1 | - | 19 | 2593 | 9056978 | ||
BX951442.1 | - | 3 | 247 |