Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002580.2 | NP_002571.1 | 4 | 347 | ||
NM_138937.2 | NP_620354.1 | 4 | 324 | |||
NM_138938.2 | NP_620355.1 | 3 | 634 | |||
GenBank mRNA | AK291973.1 | - | 4 | 323 | ||
AK311526.1 | - | 4 | 856 | |||
AY544128.1 | - | 3 | 371 | |||
AY544130.1 | - | 3 | 317 | |||
BC036776.1 | - | 3 | 328 | 5749980 | ||
BQ777112.1 | - | 4 | 347 | |||
D13510.1 | - | 3 | 323 | |||
M84337.1 | - | 3 | 328 | |||
S51768.1 | - | 3 | 328 | |||
X68641.1 | - | 3 | 299 |