Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002580.2 | NP_002571.1 | 6 | 700 | ||
NM_138937.2 | NP_620354.1 | 6 | 677 | |||
NM_138938.2 | NP_620355.1 | 5 | 987 | |||
GenBank mRNA | BC036776.1 | - | 5 | 681 | 5749980 | |
D13510.1 | - | 5 | 676 | |||
M84337.1 | - | 5 | 682 | |||
S51768.1 | - | 5 | 681 | |||
X68641.1 | - | 5 | 652 |