Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002617.3 | NP_002608.1 | 4 | 705 | ||
NM_153818.1 | NP_722540.1 | 4 | 765 | |||
XM_011541573.1 | XP_011539875.1 | 4 | 813 | |||
GenBank mRNA | AB013818.1 | - | 4 | 705 | ||
AF060502.1 | - | 4 | 668 | |||
AK124816.1 | - | 4 | 683 | |||
BC000543.2 | - | 4 | 724 | 3163782 | ||
BC018198.1 | - | 4 | 687 | 3882695 | ||
CR542077.1 | - | 4 | 636 |