Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145252.1 | NP_001138724.1 | 7 | 1190 | ||
NM_002621.2 | NP_002612.1 | 8 | 1306 | |||
XM_017029575.1 | XP_016885064.1 | 6 | 1812 | |||
GenBank mRNA | AK122955.1 | - | 6 | 2362 | ||
AK310695.1 | - | 7 | 1185 | |||
BC015756.1 | - | 7 | 1124 | 4850121 | ||
KU178249.1 | - | 7 | 1064 | |||
KU178250.1 | - | 8 | 994 | |||
M83652.1 | - | 7 | 1136 | |||
X57748.1 | - | 7 | 1129 |