Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145252.1 | NP_001138724.1 | 9 | 1479 | ||
NM_002621.2 | NP_002612.1 | 10 | 1595 | |||
XM_017029575.1 | XP_016885064.1 | 8 | 2101 | |||
GenBank mRNA | BC015756.1 | - | 9 | 1413 | 4850121 | |
KU178249.1 | - | 9 | 1353 | |||
KU178250.1 | - | 10 | 1283 | |||
M83652.1 | - | 9 | 1425 | |||
X57748.1 | - | 9 | 1418 |