Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031848.1 | NP_001027018.1 | 4, 5 | 566 | ||
NM_001276490.1 | NP_001263419.1 | 3, 4 | 388 | |||
NM_002640.3 | NP_002631.3 | 4, 5 | 566 | |||
NM_198833.1 | NP_942130.1 | 4, 5 | 583 | |||
XM_006722484.2 | XP_006722547.1 | 4, 5 | 514 | |||
XM_011526025.1 | XP_011524327.1 | 4, 5 | 567 | |||
XM_017025790.1 | XP_016881279.1 | 4, 5 | 567 | |||
XM_017025791.1 | XP_016881280.1 | 4, 5 | 511 | |||
GenBank mRNA | AK313782.1 | - | 4, 5 | 483 | ||
AL708543.1 | - | 4, 5 | 531 | |||
BC034528.1 | - | 4, 5 | 476 | 4780455 | ||
BQ435307.1 | - | 4, 5 | 589 | |||
BX571754.1 | - | 4, 5 | 474 | |||
DC384755.1 | - | 3, 4 | 322 | |||
L40377.1 | - | 4, 5 | 497 | |||
U92984.1 | - | 4, 5 | 475 |