Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002643.3 | NP_002634.1 | 2, 3 | 382 | ||
NM_173074.2 | NP_775097.1 | 2, 3 | 382 | |||
XM_005264369.2 | XP_005264426.1 | 2, 3 | 317 | |||
XM_011532908.2 | XP_011531210.1 | 2, 3 | 317 | |||
GenBank mRNA | AK291108.1 | - | 2, 3 | 337 | ||
BC021725.1 | - | 2, 3 | 322 | 5269689 | ||
BC029408.1 | - | 2, 3 | 335 | 4664848 | ||
CR457000.1 | - | 1, 2 | 218 | |||
D13435.1 | - | 2, 3 | 285 |