Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006221.3 | NP_006212.1 | 2 | 262 | ||
NR_038422.2 | 3 | 315 | ||||
NR_038830.1 | 3 | 315 | ||||
XM_011528068.2 | XP_011526370.1 | 4 | 253 | |||
GenBank mRNA | AF086512.1 | - | 2 | 129 | ||
AK092970.1 | - | 1 | 2274 | |||
AK097558.1 | - | 2 | 146 | |||
AK291074.1 | - | 2 | 146 | |||
BC002899.1 | - | 2 | 151 | 3941595 | ||
BC031971.1 | - | 3 | 1159 | |||
BG912418.1 | - | 3 | 182 | |||
BI827520.1 | - | 2 | 267 | |||
BI827672.1 | - | 2 | 264 | |||
BT019331.1 | - | 2 | 124 | |||
CD675162.1 | - | 3 | 175 | |||
CR407654.1 | - | 2 | 124 | |||
U49070.1 | - | 2 | 148 |