Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000930.4 | NP_000921.1 | 13 | 1650 | ||
NM_001319189.1 | NP_001306118.1 | 11 | 1383 | |||
NM_033011.3 | NP_127509.1 | 12 | 1512 | |||
GenBank mRNA | A03776.1 | - | 12 | 1427 | ||
A07197.1 | - | 12 | 1417 | |||
AF260825.1 | - | 7 | 892 | |||
AK289387.1 | - | 13 | 1602 | |||
AK290575.1 | - | 13 | 1602 | |||
AK297738.1 | - | 11 | 1224 | |||
AK297937.1 | - | 10 | 1113 | |||
AK299205.1 | - | 13 | 1492 | |||
AK300984.1 | - | 13 | 1302 | |||
AK313342.1 | - | 12 | 1353 | |||
AY221101.1 | - | 12 | 1393 | |||
BC002795.2 | - | 12 | 1338 | 3618149 | ||
BC007231.1 | - | 13 | 1474 | 3161005 | ||
BC013968.2 | - | 13 | 1476 | 3163742 | ||
BC018636.2 | - | 13 | 1476 | 3637273 | ||
BC095403.1 | - | 13 | 1491 | 30528441 | ||
BT007060.1 | - | 12 | 1393 | |||
D01096.1 | - | 11 | 1305 | |||
FM956488.1 | - | 12 | 1404 | |||
M15518.1 | - | 13 | 1469 | |||
M18182.1 | - | 12 | 1406 | |||
X02901.1 | - | 12 | 1410 | |||
X07393.1 | - | 13 | 1446 | |||
X13097.1 | - | 10 | 987 |