Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000930.4 | NP_000921.1 | 5 | 604 | ||
NM_001319189.1 | NP_001306118.1 | 5 | 604 | |||
NM_033011.3 | NP_127509.1 | 4 | 466 | |||
GenBank mRNA | A03776.1 | - | 4 | 381 | ||
A07197.1 | - | 4 | 371 | |||
AK289387.1 | - | 5 | 556 | |||
AK290575.1 | - | 5 | 556 | |||
AK297738.1 | - | 5 | 445 | |||
AK299205.1 | - | 5 | 446 | |||
AK300984.1 | - | 5 | 445 | |||
AK313342.1 | - | 4 | 307 | |||
AY221101.1 | - | 4 | 347 | |||
BC002795.2 | - | 4 | 292 | 3618149 | ||
BC007231.1 | - | 5 | 428 | 3161005 | ||
BC013968.2 | - | 5 | 430 | 3163742 | ||
BC018636.2 | - | 5 | 430 | 3637273 | ||
BC095403.1 | - | 5 | 445 | 30528441 | ||
BT007060.1 | - | 4 | 347 | |||
BX641021.1 | - | 1 | 567 | |||
D01096.1 | - | 3 | 259 | |||
DA549782.1 | - | 5 | 556 | |||
FM956488.1 | - | 4 | 358 | |||
M15518.1 | - | 5 | 423 | |||
M18182.1 | - | 4 | 360 | |||
X02901.1 | - | 4 | 364 | |||
X07393.1 | - | 5 | 400 | |||
X13097.1 | - | 5 | 455 |