Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278717.1 | NP_001265646.1 | 2, 3 | 305 | ||
NM_001278718.1 | NP_001265647.1 | 2, 3 | 199 | |||
NM_005031.4 | NP_005022.2 | 2, 3 | 195 | |||
NM_021902.3 | NP_068702.1 | 2, 3 | 171 | |||
XM_017026874.1 | XP_016882363.1 | 2, 3 | 150 | |||
XM_017026875.1 | XP_016882364.1 | 2, 3 | 365 | |||
XM_017026876.1 | XP_016882365.1 | 2, 3 | 300 | |||
GenBank mRNA | AA524547.1 | - | 6, 7 | 384 | ||
AK289811.1 | - | 2, 3 | 138 | |||
BC032800.2 | - | 1, 2 | 65 | 5179163 | ||
BM691130.1 | - | 2, 3 | 138 | |||
BM856555.1 | - | 2, 3 | 132 | |||
BX115036.1 | - | 2, 3 | 195 | |||
GQ258667.1 | - | 1, 2 | 43 | |||
H57207.1 | - | 2, 3 | 195 | |||
HY107209.1 | - | 2, 3 | 172 | |||
HY111815.1 | - | 2, 3 | 199 | |||
HY151101.1 | - | 2, 3 | 160 | |||
HY151827.1 | - | 2, 3 | 307 | |||
R46545.1 | - | 2, 3 | 212 | |||
R72515.1 | - | 2, 3 | 135 | |||
U72245.1 | - | 1, 2 | 61 |