Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278717.1 | NP_001265646.1 | 6 | 468 | ||
NM_001278718.1 | NP_001265647.1 | 6 | 362 | |||
NM_005031.4 | NP_005022.2 | 6 | 358 | |||
NM_021902.3 | NP_068702.1 | 6 | 334 | |||
XM_017026874.1 | XP_016882363.1 | 6 | 313 | |||
XM_017026875.1 | XP_016882364.1 | 6 | 528 | |||
XM_017026876.1 | XP_016882365.1 | 6 | 463 | |||
GenBank mRNA | AA524547.1 | - | 3 | 221 | ||
AK289811.1 | - | 6 | 301 | |||
BC032800.2 | - | 5 | 228 | 5179163 | ||
BM691130.1 | - | 6 | 301 | |||
BM856555.1 | - | 6 | 295 | |||
BX115036.1 | - | 6 | 358 | |||
HY111815.1 | - | 6 | 362 | |||
HY151101.1 | - | 6 | 323 | |||
R72515.1 | - | 6 | 298 | |||
U72245.1 | - | 5 | 224 |