Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136007.1 | NP_001129479.1 | 6, 7 | 511 | ||
NM_001136008.1 | NP_001129480.1 | 4, 5 | 305 | |||
NM_001136009.1 | NP_001129481.1 | 5, 6 | 389 | |||
NM_001136010.1 | NP_001129482.1 | 4, 5 | 305 | |||
NM_001136011.1 | NP_001129483.1 | 5, 6 | 389 | |||
NM_001136012.1 | NP_001129484.1 | 4, 5 | 305 | |||
NM_005971.3 | NP_005962.1 | 4, 5 | 305 | |||
NM_021910.2 | NP_068710.1 | 5, 6 | 389 | |||
XM_005258996.4 | XP_005259053.1 | 3, 4 | 210 | |||
XM_011527032.2 | XP_011525334.1 | 6, 7 | 518 | |||
XM_017026877.1 | XP_016882366.1 | 4, 5 | 255 | |||
XM_017026878.1 | XP_016882367.1 | 3, 4 | 175 | |||
GenBank mRNA | AK311689.1 | - | 4, 5 | 204 | ||
BC005238.1 | - | 3, 4 | 119 | 3997821 | ||
BC090044.1 | - | 4, 5 | 254 | 30395606 | ||
BE793135.1 | - | 2, 3 | 79 | |||
BI715233.1 | - | 3, 4 | 95 | |||
BM549935.1 | - | 2, 3 | 117 | |||
BQ676323.1 | - | 5, 6 | 337 | |||
BT006712.1 | - | 2, 3 | 63 | |||
BU185141.1 | - | 4, 5 | 192 | |||
CA948888.1 | - | 1, 2 | 32 | |||
CB128589.1 | - | 3, 4 | 118 | |||
CR456945.1 | - | 2, 3 | 63 | |||
CR542197.1 | - | 2, 3 | 63 | |||
DA438143.1 | - | 3, 4 | 119 | |||
DA890604.1 | - | 3, 4 | 147 | |||
DB202948.1 | - | 3, 4 | 120 | |||
U28249.1 | - | 5, 6 | 322 | |||
X93036.1 | - | 3, 4 | 122 |