Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002674.3 | NP_002665.2 | 1 | 295 | ||
XM_017019483.1 | XP_016874972.1 | 1 | 1990 | |||
GenBank mRNA | BC018048.1 | - | 1 | 283 | 4794545 | |
CB047052.1 | - | 3 | 492 | |||
CB047053.1 | - | 1 | 304 | |||
M57703.1 | - | 1 | 274 |