Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMCHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003922.1 | 3 | 1603 | |||
GenBank mRNA | AY008412.1 | - | 3 | 1603 | ||
AY008413.1 | - | 3 | 1603 | |||
EU921425.1 | - | 4 | 398 | |||
EU921431.1 | - | 4 | 315 | |||
EU921433.1 | - | 4 | 350 |