Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032940.2 | NP_116558.1 | 1, 2 | 174 | ||
GenBank mRNA | AF008443.1 | - | 1, 2 | 88 | ||
AJ224143.1 | - | 1, 2 | 127 | |||
AJ224144.1 | - | 1, 2 | 127 | |||
AK293112.1 | - | 1, 2 | 132 | |||
AK295582.1 | - | 1, 2 | 153 | |||
AK300124.1 | - | 145 | ||||
BC000409.2 | - | 1, 2 | 105 | 2960943 | ||
BC003159.1 | - | 1, 2 | 146 | 2957867 | ||
BC028157.1 | - | 1, 2 | 130 | 5240953 | ||
BI458170.1 | - | 1, 2 | 182 | |||
CR542041.1 | - | 1, 2 | 88 | |||
J05448.1 | - | 1, 2 | 129 |