Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008709.1 | NP_001008709.1 | 1 | 138 | ||
NM_002708.3 | NP_002699.1 | 1 | 138 | |||
NM_206873.1 | NP_996756.1 | 1 | 138 | |||
GenBank mRNA | AK313586.1 | - | 1 | 47 | ||
AL557336.3 | - | 1 | 162 | |||
BC001888.1 | - | 1 | 59 | 3546588 | ||
BC004482.2 | - | 1 | 79 | 3689198 | ||
BC008010.1 | - | 1 | 79 | 3528742 | ||
BE278068.1 | - | 1 | 58 | |||
BE793459.1 | - | 1 | 140 | |||
BG480286.1 | - | 1 | 138 | |||
BM799599.1 | - | 1 | 77 | |||
BT006629.1 | - | 1 | 15 | |||
M63960.1 | - | 1 | 44 | |||
S57501.1 | - | 1 | 26 | |||
X70848.1 | - | 1 | 42 |