Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006741.3 | NP_006732.3 | 4 | 403 | ||
XM_005268995.4 | XP_005269052.1 | 4 | 383 | |||
GenBank mRNA | AY063766.1 | - | 4 | 234 | ||
BC022470.1 | - | 4 | 403 | 4819907 | ||
BP211831.1 | - | 4 | 403 | |||
CR457003.1 | - | 4 | 232 | |||
U48707.1 | - | 4 | 240 |